Mdivi -1 ≥97%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: Mdivi-1, 3-(2,4-Dichloro-5-methoxyphenyl)-2-mercaptoquinazolin-4(3H)-one, 3-(2,4-Dichloro-5-methoxyphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

BMLCM1270010 BMLCM1270050
ENZOBMLCM1270010EA 1225 SEK
ENZOBMLCM1270010 ENZOBMLCM1270050
Mdivi -1 ≥97%
Mdivi -1

Mdivi-1 is a selective inhibitor of mitochondrial division in yeast and mammalian cells which acts via inhibiting the mitochondrial division dynamin. In cells, Mdivi-1 inhibits apoptosis by inhibiting mitochondrial outer membrane permeabilization. Mdivi-1 causes the rapid (50= ~10µM. In yeast, time-lapse fluorescence microscopy revealed no detectable mitochondrial division after treatment with Mdivi-1. It inhibits Dnm1 GTPase activity in a dose dependent manner with an estimated IC50 of 1-10µM but is not a general inhibitor of GTPases.


  • Inhibitor of mitochondrial division


White solid.


Soluble in DMSO (25mg/ml).

Formel: C₁₅H₁₀Cl₂N₂O₂S
MW: 353,2 g/mol
Storage Temperature: Frys
MDL Number: MFCD00974506
CAS nummer: 338967-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4688b457560944b7e382e87d17432f4fc20c55f0&vid=95aa8482089ea2c49a6d34947af4638c96dc8924&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR