XAV939 ≥98%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms:
2-(4-(Trifluoromethyl)phenyl)-3,5,7,8-tetrahydro-4H-thiopyrano[4,3-d]pyrimidin-4-one, XAV-939
BMLWN1000005
BMLWN1000025
ENZOBMLWN1000005EA
735
SEK
ENZOBMLWN1000005
ENZOBMLWN1000025
XAV939 ≥98%
XAV939
PARP5a inhibitor
Potent inhibitor of Tankyrase 1 (IC50=11nM) and Tankyrase 2 (IC50=4nM). Inhibition of tankyrase promotes stabilization of axin resulting in β-catenin degradation and Wnt signalling disruption. Inhibits proliferation of the μ-catenin-dependent colon carcinoma cell line DLD-1. Promotes cardiomyogenic development in mesoderm progenitor cells.
Formel:
C₁₄H₁₁F₃N₂OS MW: 312,3102 g/mol |
MDL Number:
MFCD16879017 CAS nummer: 284028-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...