H-7 dihydrochloride ≥98%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms: 5-((2-Methylpiperazin-1-yl)sulphonyl)isoquinoline dihydrochloride, Protein kinase inhibitor H-7 2HCl
BMLEI1480010 BMLEI1480050
ENZOBMLEI1480010EA 1225 SEK
ENZOBMLEI1480010 ENZOBMLEI1480050
H-7 dihydrochloride ≥98%
H-7 dihydrochloride
Inhibits PKA (Ki=3.0μM), PKG (Ki=5.8μM), MLCK (Ki=97μM) and PKC (Ki=6μM). Also inhibits myosin light chain kinase. Induces apoptotic DNA fragmentation and cell death. Inhibits telomerase activity in treated NPC-076 cells. Cell permeable.
  • PKC inhibitor
White to light yellow solid.
Soluble in water (50mg/ml), 100% ethanol or DMSO.
Formel: C₁₄H₁₇N₃O₂S·2HCl
MW: 364,29 g/mol
Smältpunkt: 219…222 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00036961
CAS nummer: 108930-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5f2af9ad323b668b4907d910e9e6bc88d3e4852&vid=54ddb51a517f236c3ff609281177431a76bc8d8b&prid=web&clid=SAPDAM