Zolpidemphenyl-4-carboxylic acid ≥98%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: Zolpidem Metabolite I, 4-[3-[2-(Dimethylamino)-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl] benzoic acid

BMLJ1360001 BMLJ1360005
ENZOBMLJ1360001EA 5150 SEK
ENZOBMLJ1360001 ENZOBMLJ1360005
Zolpidemphenyl-4-carboxylic acid ≥98%
Zolpidemphenyl-4-carboxylic acid

Major metabolite of zolpidem.


Zolpidem metabolite I is the major metabolite of zolpidem. Zolpidem is a non-benzodiazepine compound that acts as an agonist to the GABA-Aγ1 subunit, which has been shown to treat insomia. Intake of zolpidem can be detected by quantification of the metabolites produced. Zolpidem metabolite I shows two absorption peaks in methanol. It emits blue fluorescence upon exiting at 303 nm in water (λem= 373 nm).


Soluble in methanol, DMF and DMSO. Slightly soluble in water and ethyl acetate.

Formel: C₁₉H₁₉N₃O₃
MW: 337,37 g/mol
Storage Temperature: Frys
CAS nummer: 109461-65-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd5179fa22ef87ab221cfdc3b114689897f4c749&vid=a6afb3da03a275efd7e04baa9da9bf3ab526eeb6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity ≥98% (HPLC)
Identity Determined by MS and H-NMR
Absorbance maximum 247 nm and 303 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR