PD-0173074 ≥99%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: PD 173074, 1-(tert-Butyl)-3-[7-[[4-(diethylamino)butyl]amino]-3-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-2-yl]urea, PD173074, 1-(tert-Butyl)-3-(2-((4-(diethylamino)butyl)amino)-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl)urea

ENZCHM1120005 ENZCHM1120010
ENZOENZCHM1120005EA 2875 SEK
ENZOENZCHM1120005 ENZOENZCHM1120010
PD-0173074 ≥99%
PD-0173074

Cell-permeable, potent inhibitor of FGF and VEGF receptors.


A cell-permeable compound that acts as a potent, ATP-competitive, and reversible inhibitor of FGF and VEGF receptors. PD 173074 inhibits PDGFR and c-Src only at much higher concentration. Shown to inhibit the autophosphorylation of endogenous FGFR1 and overexpressed VEGFR2.

Formel: C₂₈H₄₁N₇O₃
MW: 523,7 g/mol
MDL Number: MFCD08705327
CAS nummer: 219580-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1bcb8a23701f1ad7dd3f222808b5312ac4a465a&vid=64d60247924281721ff0ff9bad1d97999befef1a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR