AZD-8055 ≥98%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: (5-(2,4-Bis((S)-3-methylmorpholino)pyrido[2,3-d]pyrimidin-7-yl)-2-methoxyphenyl)methanol, CCG168

ENZCHM1930005 ENZCHM1930010
ENZOENZCHM1930005EA 1075 SEK
ENZOENZCHM1930005 ENZOENZCHM1930010
AZD-8055 ≥98%
AZD-8055

Inhibitor of mTOR kinase selective against PI3K


AZD8055 is a novel ATP-competitive inhibitor of mTOR kinase activity with an IC50 of 0.8 nM. It shows excellent selectivity (~1,000-fold) against all class I phosphatidylinositol 3-kinase (PI3K) isoforms and other members of the PI3K-like kinase family.

Formel: C₂₅H₃₁N₅O₄
MW: 465,5 g/mol
MDL Number: MFCD16660191
CAS nummer: 1009298-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c10d707a9276adb44c78571bfaacc47c4e2b3ed8&vid=526af5801c6f3d9bdf1698fa812d0d70fe4342fb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR