MK-2206 dihydrochloride ≥99%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms: 8-(4-(1-Aminocyclobutyl)phenyl)-9-phenyl-[1,2,4]triazolo[3,4-f][1,6]naphthyridin-3(2H)-one dihydrochloride, MK-2206 2HCl
ENZCHM1640005 ENZCHM1640010
ENZOENZCHM1640005EA 1575 SEK
ENZOENZCHM1640005 ENZOENZCHM1640010
MK-2206 dihydrochloride ≥99%
MK-2206 dihydrochloride
Highly selective inhibitor of Akt1, Akt2 and Akt3
MK-2206 is a highly selective inhibitor of Akt1, Akt2 and Akt3 with IC50 of 8 nM, 12 nM and 65 nM, respectively.
Formel: C₂₅H₂₁N₅O·2HCl
MW: 480,4 g/mol
MDL Number: MFCD14584463
CAS nummer: 1032350-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=037240b268f1cf27409fe63a5748de534e3e7075&vid=fed7611b1217275a6a7227f9a3f0ded2b72aab7c&prid=web&clid=SAPDAM