SB216763 ≥98%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: 3-(2,4-Dichlorophenyl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione, SB-216763

BMLEI3120001 BMLEI3120005
ENZOBMLEI3120001EA 945 SEK
ENZOBMLEI3120001 ENZOBMLEI3120005
SB216763 ≥98%
SB216763

Potent and selective inhibitor of GSK-3β (IC50=34nM). Stimulates glycogen synthesis in human liver cells and mimics other actions of insulin. Protects primary neurons from death induced by PI-3 kinase pathway inhibition suggesting therapeutic potential in neurodegenerative diseases and stroke. Reduces pulmonary inflammation and fibrosis in a mouse model.


  • GSK inhibitor


Orange solid


Soluble in DMSO (>25mg/ml).

Formel: C₁₉H₁₂Cl₂N₂O₂
MW: 371,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09753369
CAS nummer: 280744-09-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a5093345a53372e4c0f30dbdc20f5510b6689bb&vid=acbc5cdc0b10b595202cdc50c6f04e5b78b9745d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR