Momelotinib ≥97%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms:
N-(Cyanomethyl)-4-(2-((4-morpholinophenyl)amino)pyrimidin-4-yl)benzamide, CYT387
ENZCHM1970001
ENZCHM1970005
ENZOENZCHM1970001EA
795
SEK
ENZOENZCHM1970001
ENZOENZCHM1970005
Momelotinib ≥97%
Momelotinib
Selective inhibitor of JAK1 and JAK2 kinases including the JAK2 V617F mutant
Using ATP kinase assays, the IC50 values for CYT387 are a follows - JAK1 (11nM), JAK2, (18nM), JAK2 V617F (23nM) JAK3 (155nM), and TYK2 (17nM). In testing kinase arrays, CYT387 also had activity (defined as an IC50 < 100nM) detected against CDK2/cyclin A, MAPK8 (JNK1), PRKCN (PKD3), PRKD1 (PKCμ), ROCK2, and TBK1 kinase as well.
Formel:
C₂₃H₂₂N₆O₂ MW: 414,5 g/mol |
MDL Number:
MFCD16038899 CAS nummer: 1056634-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...