(R)-BI-2536 ≥97%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: BI-2536, BI 2536, (R)-4-((8-Cyclopentyl-7-ethyl-5-methyl-6-oxo-5,6,7,8-tetrahydropteridin-2-yl)amino)-3-methoxy-N-(1-methylpiperidin-4-yl)benzamide

ENZCHM1600005 ENZCHM1600010
ENZOENZCHM1600005EA 1800 SEK
ENZOENZCHM1600005 ENZOENZCHM1600010
(R)-BI-2536 ≥97%
(R)-BI-2536

Potent and selective inhibitor of mammalian Plk1


Polo-like kinase 1 (Plk1) is a regulator of the cell cycle that has been implicated in the pathology of many cancers. BI-2536 is a potent and selective small-molecule inhibitor of mammalian Plk1. It has inhibitory activity at subnanomolar concentrations and inhibits tumor growth in multiple tumor lines with IC50 values below 1 µM. BI-2536 also shows an >1,000-fold selectivity for PIK1 versus a large panel of other kinases. BI-2536 also demonstrated low Kd values against sister kinases Plk2 and Plk3. Preclinical studies in human cancer cell lines have shown that BI 2536 disrupts spindle assembly, resulting in mitotic arrest and inducing apoptosis. BI-2536 is currently in clinical trials against several types of solid tumor cancers.

Formel: C₂₈H₃₉N₇O₃
MW: 521,7 g/mol
MDL Number: MFCD10565924
CAS nummer: 755038-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfeae6a9dd2f75adfcc297f5f0c8b8e0c0208e3f&vid=6804b1a6570069c8141e1fa7dcf749381f543f4d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR