Tricyclodecan-9-yl xanthogenate potassium salt ≥98%
Leverantör: ENZO LIFE SCIENCES AG
Selective inhibitor of phosphatidylcholine-specific phospholipase C (PC-PLC). Has been used to study the coupling of phosphatidylcholine-specific phospholipase C with sphingomyelinase. Shows antitumor and antiviral activity. Inhibits basement membrane collagen synthesis, which is the last step in the development of a new blood vessel. Inhibits induction of nitric oxide synthases (NOS). Induces apoptosis. It indirectly inhibits phorbol ester stimulated PLD activity.
- Phospholipase inhibitor
Off-white solid.
Soluble in water (25mg/ml).
Formel:
C₁₁H₁₅KOS₂ MW: 266,5 g/mol Storage Temperature: Frys |
MDL Number:
MFCD00171401 CAS nummer: 83373-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...