Bisindolylmaleimide I hydrochloride ≥99%
Leverantör: ENZO LIFE SCIENCES AG
ENZCHM1080001
ENZCHM1080005
ENZOENZCHM1080001EA
785
SEK
ENZOENZCHM1080001
ENZOENZCHM1080005
Bisindolylmaleimide I hydrochloride ≥99%
Bisindolylmaleimide I hydrochloride
Anti-inflammatory agent and protein kinase C inhibitor.
Enhanced, water soluble form of Bisindolylmaleimide I.
Formel:
C₂₅H₂₄N₄O₂·HCl MW: 452,5·36,5 g/mol |
CAS nummer:
176504-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...