P32/98 ≥98%
Leverantör: ENZO LIFE SCIENCES AG
BMLPI1420010
BMLPI1420050
ENZOBMLPI1420010EA
1750
SEK
ENZOBMLPI1420010
ENZOBMLPI1420050
P32/98 ≥98%
P32/98
DPPIV/DPP4 inhibitor
P32/98 (Ile-thiazolidide) is a specific, competitive transition-state substrate analog inhibitor of dipeptidyl peptidase IV (DPIV; DPPIV; CD26), with a Ki of 130 nM. Useful in diabetes research. Has been used in vivo and in tissue culture.
Formel:
C₁₃H₂₂N₂O₅S MW: 318,388 g/mol |
MDL Number:
MFCD22741399 CAS nummer: 136259-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...