Fumagillin ≥95%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: (2E,4E,6E,8E)-10-(((3R,4S,5S,6R)-5-methoxy-4-((2R,3R)-2-methyl-3-(3-methylbut-2-en-1-yl)oxiran-2-yl)-1-oxaspiro[2.5]octan-6-yl)oxy)-10-oxodeca-2,4,6,8-tetraenoic acid

BMLCT1000001 BMLCT1000005
ENZOBMLCT1000001EA 1475 SEK
ENZOBMLCT1000001 ENZOBMLCT1000005
Fumagillin ≥95%
Fumagillin

Methionine aminopeptidase 2 inhibitor


Inhibitor of angiogenesis and endothelial cell proliferation. Specific inhibitor of methionine aminopeptidase type II (MetAP-II) by covalently modifying a conserved active-site histidine. Precursor to TNP-470. Exhibits antitumor activity in estrogen-induced pituitary adenoma. Antineoplastic. Anti-infective.


Isolated from Aspergillus fumigatus.

Formel: C₂₆H₃₄O₇
MW: 458,551 g/mol
MDL Number: MFCD03990453
CAS nummer: 23110-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6e812745b76551ff3f756c5501642ed1bd43acc&vid=e82df31e482c19b5dd24d38c87c739280d32125c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR