D-erythro MAPP ≥98%

Leverantör: ENZO LIFE SCIENCES AG

BMLSL2210005 BMLSL2210025
ENZOBMLSL2210005EA 945 SEK
ENZOBMLSL2210005 ENZOBMLSL2210025
D-erythro MAPP ≥98%
D-erythro MAPP

Alkaline ceramidase inhibitor


A cell-permeable and specific inhibitor of alkaline ceramidase, which can raise intracellular ceramide levels by blocking the hydrolysis of ceramide to sphingosine and free fatty acid. Enhances the apoptotic response to ionizing radiation by enhancing the ceramide signal.

Formel: C₂₃H₃₉NO₂
MW: 361,57 g/mol
MDL Number: MFCD01076527
CAS nummer: 143492-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03bee86f1e512f352ea87307db78149cb516447d&vid=a6552b01de010423cfa57fd681244ee8f7033ac8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR