Bourgeonal ≥98%, colourless oil
Leverantör: ENZO LIFE SCIENCES AG
|
Fara
|
Synonyms:
3-(4-(tert-Butyl)phenyl)propanal
BMLN1560005
BMLN1560025
ENZOBMLN1560005EA
1350
SEK
ENZOBMLN1560005
ENZOBMLN1560025
Bourgeonal ≥98%, colourless oil
Bourgeonal
hOR17-4 ligand
A previously undescribed human testicular olfactory receptor known as hOR17-4 was cloned and functionally expressed. Bourgeonal is a potent agonist at hOR17-4 and acts as a strong chemoattractant in sperm behavioral assays. The bourgeonal, hOR17-4 signaling pathway potentially governs chemical communication between sperm and egg and may be used to manipulate or block fertilization.
Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml).
Formel:
C₁₃H₁₈O MW: 190,286 g/mol Storage Temperature: Frys |
MDL Number:
MFCD00169819 CAS nummer: 18127-01-0 |
Specifikation testresultat
Purity | ≥98% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...