Amifostine ≥98%

Leverantör: ENZO LIFE SCIENCES AG
Varning

BMLGR3410010 BMLGR3410050
ENZOBMLGR3410010EA 965 SEK
ENZOBMLGR3410010 ENZOBMLGR3410050
Amifostine ≥98%
Amifostine

HIF-1α activator.


Amifostine is a broad-spectrum radio- and chemoprotector. It restores transcriptional activity of specific p53 mutants, prevents chemotherapy-induced apoptosis and upregulates HIF-1 & alpha.


Soluble in water.

Formel: C₅H₁₅N₂O₃PS
Storage Temperature: Kylskåp
CAS nummer: 20537-88-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80e51c10a566d35bb8baec7ac4f11f7430343c5f&vid=771b42d3319d6227800ab485783c8cf15a3224e7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity ≥98% (HPLC)
Identity Determined by IR
Appearance White to off-white crystalline powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR