ZM-336372 ≥95%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms: 3-(Dimethylamino)-N-(3-(4-hydroxybenzamido)-4-methylphenyl)benzamide, ZM 336372
BMLEI2980005 BMLEI2980025
ENZOBMLEI2980005EA 1975 SEK
ENZOBMLEI2980005 ENZOBMLEI2980025
ZM-336372 ≥95%
ZM-336372
Potent and specific inhibitor of the protein kinase c-Raf (IC50=70nM). Has no significant effect on many other protein kinases tested (even at 50µM) with the exception of p38 (SAPK2a) (IC50=2µM) and SAPK2b (p38β) (IC50=2µM). Induces a paradoxical >100-fold activation of c-Raf in whole cells.
  • Raf inhibitor
White to off-white solid.
Soluble in DMSO (200mg/ml).
Formel: C₂₃H₂₃N₃O₃
MW: 389,455 g/mol
Storage Temperature: Frys
MDL Number: MFCD02683971
CAS nummer: 208260-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be3f45af687f724d237d612ce579f3274ef48ce2&vid=ee53d26b17fb0fa7c1626c901895a7945696a4af&prid=web&clid=SAPDAM