N,N,2-Trimethyl-5-nitrobenzenesulphonamide (synthetic) ≥98%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms: BRL-50481, BRL 50481
BMLPD1200010 BMLPD1200050
ENZOBMLPD1200010EA 965 SEK
ENZOBMLPD1200010 ENZOBMLPD1200050
N,N,2-Trimethyl-5-nitrobenzenesulphonamide (synthetic) ≥98%
N,N,2-Trimethyl-5-nitrobenzenesulphonamide
The first selective PDE7 inhibitor (Ki=180 nM).
  • PDE inhibitor
White to off-white solid.
Formel: C₉H₁₂N₂O₄S
MW: 244,27 g/mol
Smältpunkt: 65…67 °C
Storage Temperature: Omgivande
MDL Number: MFCD03039912
CAS nummer: 433695-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c32213fb0da3d4eeaad80fff3ea4e0e3e7dfcf7&vid=c4e4f9a2c8bb0042d082547e51d91413301d0211&prid=web&clid=SAPDAM