Lovastatin ≥98%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: 6alpha-Methylcompactin, Monacolin K, Mevinolin, (S)-(1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2-methylbutanoate

BMLG2260010 BMLG2260050
ENZOBMLG2260010EA 745 SEK
ENZOBMLG2260010 ENZOBMLG2260050
Lovastatin ≥98%
Lovastatin

HMG-CoA reductase inhibitor


Inhibits cholesterol/isoprenoid biosynthesis by inhibition of HMG-CoA reductase (Ki for acid form is 1nM) and blocks protein isoprenylation and reduces plasma cholesterol levels in humans. Causes cells to arrest early in the G1 phase. Induces apoptosis in cancer cells.

Formel: C₂₄H₃₆O₅
MW: 404,55 g/mol
MDL Number: MFCD00072164
CAS nummer: 75330-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ac7c44bb3bfcacb1f1152a60c37a2bd8c825610&vid=02e5429a451616e60503ccb6dd01b8eaa8d9641b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR