Anti-KDEL Receptor Mouse Monoclonal Antibody

Leverantör: ENZO LIFE SCIENCES
ADIVAAPT048D ADIVAAPT048F
ENZOADIVAAPT048DEA 3075 SEK
ENZOADIVAAPT048D ENZOADIVAAPT048F
Anti-KDEL Receptor Mouse Monoclonal Antibody
Antikroppar
The tetrapeptide KDEL, located at the carboxy-terminal sequences of luminal proteins, is a retrieval motif essential for the precise sorting of these proteins along the secretory pathway. KDEL proteins perform essential functions in the endoplasmic reticulum (ER) related to protein folding as well as assembly. The localization of chaperones and other soluble proteins to the ER is achieved by their continuous retrieval from post-ER compartments by the KDEL receptor (Erd2p), which is a membrane protein localized in the Golgi apparatus.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat KDEL Receptor. Detects a band of ~23kDa by Western blot.

Type: Primary
Antigen: KDEL Receptor
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR