Anti-UGT1A9 Mouse Monoclonal Antibody (CF647) [clone: UGT1A9/1229]

Leverantör: Biotium
BNC471229-500 BNC471229-100
BNC471229-500EA 6500 SEK
BNC471229-500 BNC471229-100
Anti-UGT1A9 Mouse Monoclonal Antibody (CF647) [clone: UGT1A9/1229]
Antikroppar
This antibody recognizes a protein of about 60 kDa, which is identified as human UGT1A9. It does not cross-react with the other UGT1A isoforms including UGT1A7, UGT1A8, and UGT1A10 and shows a high degree of specificity. UGT1A9 is a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This MAb binds to human hepatocytes and the majority of human hepatocellular carcinomas (HCC s). In frozen sections, it stains hepatic cells and may be used as a marker of the liver. This MAb also binds to cell preparations of hepatocellular carcinoma biopsies or cell lines.

Type: Primary
Antigen: UGT1A9
Clonality: Monoclonal
Clone: UGT1A9/1229
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR