Anti-CELA3B Mouse Monoclonal Antibody (CF594) [clone: CELA3B/1218]

Leverantör: Biotium

BNC941218-500 BNC941218-100
BNC941218-500EA 6150 SEK
BNC941218-500 BNC941218-100
Anti-CELA3B Mouse Monoclonal Antibody (CF594) [clone: CELA3B/1218]
Antikroppar
This MAb recognizes a protein of ~17 kDa, identified as CELA3B (Chymotrypsin like elastase family member 3B). Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, elastase 3B has little elastolytic activity. Like most of the human elastases, elastase 3B is secreted from the pancreas as a zymogen and, like other serine proteases such as trypsin, chymotrypsin and kallikrein; it has a digestive function in the intestine. Elastase 3B preferentially cleaves proteins after alanine residues. Elastase 3B may also function in the intestinal transport and metabolism of cholesterol. Both elastase 3A and elastase 3B have been referred to as protease E and as elastase 1, and excretion of this protein in fecal material is frequently used as a measure of pancreatic function in clinical assays.

Type: Primary
Antigen: CELA3B
Clonality: Monoclonal
Clone: CELA3B/1218
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR