Anti-Human Papillomavirus 16 Mouse Monoclonal Antibody (CF594) [clone: HPV16/1058]

Leverantör: Biotium
BNC941058-500 BNC941058-100
BNC941058-500EA 6500 SEK
BNC941058-500 BNC941058-100
Anti-Human Papillomavirus 16 Mouse Monoclonal Antibody (CF594) [clone: HPV16/1058]
Antikroppar
Reacts with a protein of 57 kDa, identified as the L1 protein of human papilloma virus type 16 (HPV-16). It is the major capsid protein of HPV-16. Infection with specific types of HPV has been associated with an increased risk of developing cervical neoplasia. HPV types 6 and 11 have been associated with relatively benign diseases such as genital warts but types 16 and 18 are strongly associated with cervical, vaginal, and vulvar malignancies. The antibody reacts very strongly with formalin-fixed, paraffin-embedded tissues containing HPV-16 or -33; very weak reactions were occasionally observed with biopsy specimens or smears containing HPV-6 or HPV-11. It cross-reacts with HPV37.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: HPV16/1058
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: HPV
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR