Anti-BCL2 Mouse Monoclonal Antibody (CF568) [clone: BCL2/782 BCL2/796]
Leverantör: Biotium
BNC681023-500 BNC681023-100
BNC681023-500EA 6500 SEK
BNC681023-500 BNC681023-100
Anti-BCL2 Mouse Monoclonal Antibody (CF568) [clone: BCL2/782 BCL2/796]
Antikroppar
This antibody recognizes a protein of 25-26 kDa, identified as the bcl-2 α oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 α protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicular lymphomas that express bcl-2 protein and the small number in which the neoplastic cells are bcl-2 negative.
Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: BCL2/782 BCL2/796
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human