Anti-BCL2 Mouse Monoclonal Antibody [clone: BCL2/782 BCL2/796]
Leverantör: Biotium
BNUB1023-500 BNUB1023-100
BNUB1023-500EA 5925 SEK
BNUB1023-500 BNUB1023-100
Anti-BCL2 Mouse Monoclonal Antibody [clone: BCL2/782 BCL2/796]
Antikroppar
This antibody recognizes a protein of 25-26 kDa, identified as the bcl-2 α oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 α protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicular lymphomas that express bcl-2 protein and the small number in which the neoplastic cells are bcl-2 negative.
  • Purified
Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: BCL2/782 BCL2/796
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human