Anti-KIT Mouse Monoclonal Antibody (Purified) [clone: KIT/982]
Leverantör: Biotium
BNUB0982-500 BNUB0982-100
BNUB0982-500EA 5925 SEK
BNUB0982-500 BNUB0982-100
Anti-KIT Mouse Monoclonal Antibody (Purified) [clone: KIT/982]
Antikroppar
This MAb recognizes a protein of 145 kDa, identified as CD117/p145kit. It is found on a wide variety of tumor cells including follicular and papillary carcinoma of thyroid, adenocarcinomas from endometrium, lung, ovary, pancreas, and breast as well as malignant melanoma, endodermal sinus tumor, and small cell carcinoma. However, anti-CD117 has been particularly useful in differentiating gastrointestinal stromal tumors from Kaposi's sarcoma, tumors of smooth muscle origin, fibromatosis, and neural tumors of the GI tract. Anti-CD117 is also useful in recognizing myeloblasts in bone marrow biopsy and clot section.
Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: KIT/982
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human