Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF594) [clone: AE-1]

Leverantör: Biotium
BNC940905-500 BNC940905-100
BNC940905-500EA 6500 SEK
BNC940905-500 BNC940905-100
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF594) [clone: AE-1]
Antikroppar
This MAb recognizes a 60 kDa antigen associated with the mitochondria in human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in human cells only. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignant cells and may be used to stain mitochondria of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations. This MAb is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells, including neurons and embryonic stem cells.

Type: Primary
Antigen: Mitochondrial Marker
Clonality: Monoclonal
Clone: AE-1
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR