Anti-P504S Mouse Monoclonal Antibody [clone: 13H4]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB438-05 MOB438
DIABMOB438-05EA 0 SEK
DIABMOB438-05 DIABMOB438
Anti-P504S Mouse Monoclonal Antibody [clone: 13H4]
Antikroppar
Type:
Antigen: P504S
Clonality: Monoclonal
Clone: 13H4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR