Anti-MMP2 Mouse Monoclonal Antibody [clone: A-Gel VC2]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB312 MOB312-05
DIABMOB312EA 0 SEK
DIABMOB312 DIABMOB312-05
Anti-MMP2 Mouse Monoclonal Antibody [clone: A-Gel VC2]
Antikroppar
Type:
Antigen: MMP2
Clonality: Monoclonal
Clone: A-Gel VC2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR