Anti-Microphthalmia Mouse Monoclonal Antibody [clone: C5 + D5]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB351-05 MOB351
DIABMOB351-05EA 0 SEK
DIABMOB351-05 DIABMOB351
Anti-Microphthalmia Mouse Monoclonal Antibody [clone: C5 + D5]
Antikroppar
Type:
Antigen: Microphthalmia
Clonality: Monoclonal
Clone: C5 + D5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR