Anti-CDC25B Mouse Monoclonal Antibody [clone: DCS-162]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB338 MOB338-05
DIABMOB338EA 0 SEK
DIABMOB338 DIABMOB338-05
Anti-CDC25B Mouse Monoclonal Antibody [clone: DCS-162]
Antikroppar
Type:
Antigen: CDC25B
Clonality: Monoclonal
Clone: DCS-162
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR