Anti-CD15 Mouse Monoclonal Antibody [clone: MY-1]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB024-05 MOB024
DIABMOB024-05EA 0 SEK
DIABMOB024-05 DIABMOB024
Anti-CD15 Mouse Monoclonal Antibody [clone: MY-1]
Antikroppar
Type:
Antigen: CD15
Clonality: Monoclonal
Clone: MY-1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR