Anti-CH Mouse Monoclonal Antibody [clone: CALP]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB345 MOB345-05
DIABMOB345EA 0 SEK
DIABMOB345 DIABMOB345-05
Anti-CH Mouse Monoclonal Antibody [clone: CALP]
Antikroppar
Type:
Antigen: CH
Clonality: Monoclonal
Clone: CALP
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR