Anti-BCL10 Mouse Monoclonal Antibody () [clone: 151]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB442-05 MOB442
DIABMOB442-05EA 0 SEK
DIABMOB442-05 DIABMOB442
Anti-BCL10 Mouse Monoclonal Antibody () [clone: 151]
Antikroppar
Type:
Antigen: BCL10
Clonality: Monoclonal
Clone: 151
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR