Anti-GST (Pi) Mouse Monoclonal Antibody [clone: 353-10]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB065 MOB065-05
DIABMOB065EA 0 SEK
DIABMOB065 DIABMOB065-05
Anti-GST (Pi) Mouse Monoclonal Antibody [clone: 353-10]
Antikroppar
Type:
Antigen: GST (Pi)
Clonality: Monoclonal
Clone: 353-10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR