Anti-GAL3 Mouse Monoclonal Antibody [clone: 9C4]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB405-05 MOB405
DIABMOB405-05EA 0 SEK
DIABMOB405-05 DIABMOB405
Anti-GAL3 Mouse Monoclonal Antibody [clone: 9C4]
Antikroppar
Type:
Antigen: Galectin 3
Clonality: Monoclonal
Clone: 9C4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR