Anti-CK13 Mouse Monoclonal Antibody [clone: KS-1A3]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB443 MOB443-05
DIABMOB443EA 0 SEK
DIABMOB443 DIABMOB443-05
Anti-CK13 Mouse Monoclonal Antibody [clone: KS-1A3]
Antikroppar
This antibody is specific to 54 kDa protein. Cytokeratin 13 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 4. Cytokeratin 13 is expressed in squamous, non-keratinized epithelium, transitional epithelium and myoepithelium. This antibody is useful for the identification of carcinomas of trachea, sweat glands, bladder, ectocervix, tongue, esophagus, anal canal and the basal layer of keratinized epidermis.

Type:
Antigen: CK13
Clonality: Monoclonal
Clone: KS-1A3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR