Anti-COL4 Mouse Monoclonal Antibody [clone: COL-94]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB229 MOB229-05
DIABMOB229EA 0 SEK
DIABMOB229 DIABMOB229-05
Anti-COL4 Mouse Monoclonal Antibody [clone: COL-94]
Antikroppar
This antibody recognizes collagen type IV epitope located on the α1 and/or α 2 chains of human collagen type IV. This antibody shows no cross-reactivity with collagen types I, II, III, V, VI and VII and does not react with human vitronectin, fibronectin or chondroitin sulfate A, B and C.

Type:
Antigen: COL4
Clonality: Monoclonal
Clone: COL-94
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR