Anti-TYRP1 Mouse Monoclonal Antibody (CF594) [clone: TA99 TYRP1/807]
Leverantör: Biotium
BNC940808-500 BNC940808-100
BNC940808-500EA 6500 SEK
BNC940808-500 BNC940808-100
Anti-TYRP1 Mouse Monoclonal Antibody (CF594) [clone: TA99 TYRP1/807]
Antikroppar
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.
Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: TA99 TYRP1/807
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Mouse