Anti-TYRP1 Mouse Monoclonal Antibody (CF568) [clone: TYRP1/807]

Leverantör: Biotium
BNC680807-500 BNC680807-100
BNC680807-500EA 6500 SEK
BNC680807-500 BNC680807-100
Anti-TYRP1 Mouse Monoclonal Antibody (CF568) [clone: TYRP1/807]
Antikroppar
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.

Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: TYRP1/807
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR