Anti-TYRP1 Mouse Monoclonal Antibody (CF647) [clone: TA99]
Leverantör: Biotium
BNC470806-500 BNC470806-100
BNC470806-500EA 6500 SEK
BNC470806-500 BNC470806-100
Anti-TYRP1 Mouse Monoclonal Antibody (CF647) [clone: TA99]
Antikroppar
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.
Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: TA99
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Mouse