Anti-TYRP1 Mouse Monoclonal Antibody (CF405S) [clone: TA99]

Leverantör: Biotium
BNC040806-500 BNC040806-100
BNC040806-500EA 6500 SEK
BNC040806-500 BNC040806-100
Anti-TYRP1 Mouse Monoclonal Antibody (CF405S) [clone: TA99]
Antikroppar
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.

Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: TA99
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR