Anti-GFAP Mouse Monoclonal Antibody (CF640R) [clone: ASTRO/789]

Leverantör: Biotium
BNC400789-500 BNC400789-100
BNC400789-500EA 6500 SEK
BNC400789-500 BNC400789-100
Anti-GFAP Mouse Monoclonal Antibody (CF640R) [clone: ASTRO/789]
Antikroppar
This MAb recognizes a protein of ~50 kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS.

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: ASTRO/789
Conjugation:More Product Information
Order Now

SPECIFIKATIONER

Antigensymbol GFAP
Environmentally Preferable
Antikroppsnamn Glial fibrillary acidic protein
Antigen synonymer Astrocyte or Intermediate Filament Protein|Glial Fibrillary Acidic Protein (GFAP)
Antikroppstyp Primär
Klonalitet Monoclonal
Konjugation CF® 640R
Klon Clone ASTRO/789
Reaktivitet Pig,Rabbit,Chicken,Human,Bovine,Rat,Mouse
Värd Mouse
Gen-ID 514227
Isotyp IgG1 kappa
Western blot Yes
ImmunKemi Yes
ImmunoFluorescens Yes
Koncentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR