Anti-SILV Mouse Monoclonal Antibody (Purified) [clone: PMEL/783]

Leverantör: Biotium

BNUB0783-500 BNUB0783-100
BNUB0783-500EA 5625 SEK
BNUB0783-500 BNUB0783-100
Anti-SILV Mouse Monoclonal Antibody (Purified) [clone: PMEL/783]
Antikroppar
Cytotoxic T lymphocytes (CTL's) recognize melanoma-associated antigens, which belong to three main groups. These groups include tumor-associated testis-specific antigens, melanocyte differentiation antigens and mutated or aberrantly expressed antigens, which are routinely used as markers to identify melanomas based on their binding to specific monoclonal antibodies. gp100, also designated ME20-M, ME20-S and PMEL 17, is classified as a melanocyte differentiation antigen and is expressed at low levels in normal cell lines and tissues, but is upregulated in melanocytes. gp100 is a highly glycosylated protein. It is also the product of proteolytic cleavage, which results in a secreted protein.

Type: Primary
Antigen: SILV
Clonality: Monoclonal
Clone: PMEL/783
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR