Anti-BCL2 Mouse Monoclonal Antibody (CF594) [clone: BCL2/782]

Leverantör: Biotium
BNC940782-500 BNC940782-100
BNC940782-500EA 6500 SEK
BNC940782-500 BNC940782-100
Anti-BCL2 Mouse Monoclonal Antibody (CF594) [clone: BCL2/782]
Antikroppar
This antibody recognizes a protein of 25-26 kDa, identified as the bcl-2 α oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 α protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicular lymphomas that express bcl-2 protein and the small number in which the neoplastic cells are bcl-2 negative.

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: BCL2/782
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR