Anti-HSPB1 Mouse Monoclonal Antibody (CF770) [clone: HSPB1/774]
Leverantör: Biotium
BNC700774-500 BNC700774-100
BTIUBNC700774-500EA 6500 SEK
BTIUBNC700774-500 BTIUBNC700774-100
Anti-HSPB1 Mouse Monoclonal Antibody (CF770) [clone: HSPB1/774]
Antikroppar
This antibody recognizes a 24-27 kDa estrogen-regulated protein, identified as heat shock protein 27 (hsp27). Hsp27 was recently found to be identical to the estrogen-induced p29 and 24K protein. About 50% of breast carcinomas are positive for hsp27 especially those that are also positive for estrogen and/or progesterone receptor. HSP27 has also been implicated in drug resistance in cancer cells.
Type: Primary
Antigen: HSPB1
Clonality: Monoclonal
Clone: HSPB1/774
Conjugation: CF770
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Chimpanzee, Human, Mouse, Rat, Sheep