Anti-PRLR Mouse Monoclonal Antibody (CF488A) [clone: PRLR/742]

Leverantör: Biotium

BNC880742-500 BNC880742-100
BNC880742-500EA 6150 SEK
BNC880742-500 BNC880742-100
Anti-PRLR Mouse Monoclonal Antibody (CF488A) [clone: PRLR/742]
Antikroppar
This antibody recognizes a protein of 70 kDa, identified as prolactin receptor. Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor (prolactin receptor), which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.

Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: PRLR/742
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR