Anti-CHGA Mouse Monoclonal Antibody (CF488A) [clone: PHE5]

Leverantör: Biotium

BNC880461-500 BNC880461-100
BNC880461-500EA 6150 SEK
BNC880461-500 BNC880461-100
Anti-CHGA Mouse Monoclonal Antibody (CF488A) [clone: PHE5]
Antikroppar
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma. The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms. Most pituitary adenomas and prolactinomas readily express chromogranin.

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: PHE5
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR