Anti-MFGE8 Mouse Monoclonal Antibody (CF594) [clone: MFG-06]
Leverantör: Biotium
BNC940227-500 BNC940227-100
BNC940227-500EA 6500 SEK
BNC940227-500 BNC940227-100
Anti-MFGE8 Mouse Monoclonal Antibody (CF594) [clone: MFG-06]
Antikroppar
Recognizes a protein of 40-45 kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.
Type: Primary
Antigen: MFGE8
Clonality: Monoclonal
Clone: MFG-06
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human